Learn More
Medchemexpress LLC MCP-1/CCL2 Mouse H 50ug
SDP
Catalog No. 5000182445
Encompass_Preferred
MCP-1/CCL2 Protein Mouse (HEK293 His) is a cytokine belonging to the CC chemokine family that interacts with the CCR2 chemokine receptor on the cell surface to mediate inflammatory immune responses viral infections and tumorigenesis MCP-1/CCL2 Protein Mouse (HEK293 His) is a mouse MCP-1/CCL2 expressed by HEK293 with a His tag at the C-terminus[1]
Catalog No. 50-001-82445 Supplier Medchemexpress LLC Supplier No. HYP7275750UG
MCP-1/CCL2 Protein Mouse (HEK293 His) is a cytokine belonging to the CC chemokine family that interacts with the CCR2 chemokine receptor on the cell surface to mediate inflammatory immune responses viral infections and tumorigenesis MCP-1/CCL2 Protein Mouse (HEK293 His) is a mouse MCP-1/CCL2 expressed by HEK293 with a His tag at the C-terminus[1]