Learn More
Enzo Life Sciences MCP-1 monoclonal antibody (E10052) (biotin conjugate) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2810100
Encompass_Preferred
Clone: E10052. Isotype: IgG1. Immunogen: Full-length recombinant human MCP-1. Species reactivity: Human. Applications: ELISA, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing no preservative.
Catalog No. 50-201-2824