Learn More
Medchemexpress LLC HY-P7240 10ug Medchemexpress, MCP-3/CCL7 (CHO-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 502083132
Encompass_Preferred
Medchemexpress, HY-P7240 10ug MCP-3/CCL7 (CHO-expressed), Human CAS: Recombinant Human Monocyte Chemotactic Protein-3/CCL7 (CHO-expressed) is a chemotactic factor and attractant for various kinds of leukocytes, including monocytes and neutrophils. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3132 Supplier Medchemexpress LLC Supplier No. HYP724010UG
Medchemexpress, HY-P7240 10ug MCP-3/CCL7 (CHO-expressed), Human CAS: Recombinant Human Monocyte Chemotactic Protein-3/CCL7 (CHO-expressed) is a chemotactic factor and attractant for various kinds of leukocytes, including monocytes and neutrophils. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.