Learn More
Invitrogen™ Human MCP-3 ProQuantum Immunoassay Kit

Description
Specifications
Specifications
| Accession Number | P80098 |
| Assay Range | 0.064-5000 pg/mL |
| Assay Sensitivity | <0.05 pg/mL |
| Content And Storage | Contains assay dilution buffer, antibody-conjugate A, antibody-conjugate B, antibody-conjugate dilution buffer, standard, master mix, and ligase. Store at -5 to -30°C. |
| Conjugate | FAM |
| Detection Method | Taqman™ Real-time PCR |
| Immunoassay Kit Format | Proximity-based Immunoassay |
| Product Type | ProQuantum |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.