Learn More
Medchemexpress LLC HY-P7246 10ug Medchemexpress, MCP-5/CCL12 (CHO-expressed), Mouse CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP724610UG
Encompass_Preferred
Medchemexpress, HY-P7246 10ug MCP-5/CCL12 (CHO-expressed), Mouse CAS: Recombinant Mouse MCP-5/CCL12 (CHO-expressed) is a potent monocyte active chemokine that is involved in allergic inflammation and the host response to pathogens. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3134