Learn More
Genscript Corporation MCP3 CCL7 HumanCHO
Catalog No. 5000059470
Encompass
Chemokine (C-C motif) ligand 7 (CCL7) is a small cytokine that was previously called monocyte-specific chemokine 3 (MCP-3) Due to CCL7 possessing two adjacent N-terminal cysteine residues in its mature form it is classified within the subfamily of chemokines known as CC chemokines CCL7 specifically attracts monocytes and regulates macrophage function It is produced by certain tumor cell lines and by macrophages This chemokine is located on chromosome 17 in humans within a large cluster containing many other CC chemokines and is most closely related to CCL2 CCL7 can signal through the CCR1 CCR2 and CCR3 receptors
Catalog No. 50-000-59470 Supplier Genscript Corporation Supplier No. Z0323550
Chemokine (C-C motif) ligand 7 (CCL7) is a small cytokine that was previously called monocyte-specific chemokine 3 (MCP-3) Due to CCL7 possessing two adjacent N-terminal cysteine residues in its mature form it is classified within the subfamily of chemokines known as CC chemokines CCL7 specifically attracts monocytes and regulates macrophage function It is produced by certain tumor cell lines and by macrophages This chemokine is located on chromosome 17 in humans within a large cluster containing many other CC chemokines and is most closely related to CCL2 CCL7 can signal through the CCR1 CCR2 and CCR3 receptors