Learn More
AdipoGen MCPA-thioethyl
SDP
Supplier:  AdipoGen CDXM0136M500
Encompass_Preferred
Chemical. CAS 25319-90-8. Formula C11H13ClO2S. MW 244.7. Synthetic. Phenoxy herbicide. Synthetic auxin. Compound can be used as analytical reference material.
Catalog No. 50-256-0497