Learn More
Roboz Surgical MCPHEARSON VANNAS SCISSOR
SDP
Catalog No. NC9437707
Encompass
McPherson-Vannas Micro Dissecting Spring Scissors; Straight, Sharp Points; 5mm Cutting Edge; 0.15mm Tip Width; 3 in. Overall Length
Catalog No. NC9437707 Supplier Roboz Surgical Supplier No. RS5600
McPherson-Vannas Micro Dissecting Spring Scissors; Straight, Sharp Points; 5mm Cutting Edge; 0.15mm Tip Width; 3 in. Overall Length