Learn More
Roboz Surgical MCPHERSON FCP 3.5 IN ANG TYING
SDP
Catalog No. NC9457430
Encompass
McPherson Forceps; Angled; Smooth Tying Platform; 0.4 X 7mm Tip; 4 in. Length; .35mm Tip Width
Catalog No. NC9457430 Supplier Roboz Surgical Supplier No. RS5069
McPherson Forceps; Angled; Smooth Tying Platform; 0.4 X 7mm Tip; 4 in. Length; .35mm Tip Width