Learn More
World Precision Instrument McPherson Forceps, 11.5 cm (4.5 in.), Angled Tips 9.5 mm, 6.6 mm Tying Platform
SDP
Catalog No. 502538297
Encompass
McPherson forceps, utilized in surgical procedures, feature slender, angled jaws with tungsten carbide tips. The distinctive design aids in delicate tissue manipulation. Their application differs based on size, with 8.5 cm forceps offering precision for fine tasks, while 11 cm ones provide additional leverage for larger tissues. The tying platform near the base facilitates secure suturing, offering stability and control during knot tying. They are made of titanium, which offers advantages such as high strength, corrosion resistance, biocompatibility, lightweight nature, non-magnetic properties, and radiopacity. Titanium's durability ensures the forceps withstand surgical demands, and its biocompatibility supports safe interaction with body tissues. The non-magnetic property is beneficial in environments with MRI, and radiopacity aids visibility in medical imaging. They are sterilizable via autoclaving or chemical sterilant.
Catalog No. 50-253-8297 Supplier World Precision Instrument Supplier No. WP5030L
McPherson forceps, utilized in surgical procedures, feature slender, angled jaws with tungsten carbide tips. The distinctive design aids in delicate tissue manipulation. Their application differs based on size, with 8.5 cm forceps offering precision for fine tasks, while 11 cm ones provide additional leverage for larger tissues. The tying platform near the base facilitates secure suturing, offering stability and control during knot tying. They are made of titanium, which offers advantages such as high strength, corrosion resistance, biocompatibility, lightweight nature, non-magnetic properties, and radiopacity. Titanium's durability ensures the forceps withstand surgical demands, and its biocompatibility supports safe interaction with body tissues. The non-magnetic property is beneficial in environments with MRI, and radiopacity aids visibility in medical imaging. They are sterilizable via autoclaving or chemical sterilant.