Learn More
Roboz Surgical MCPHERSON STRAIGHT 3 1/2
SDP
Catalog No. NC9954786
Encompass
McPherson Forceps; Straight; Smooth Tying Platform; 0.3 X 5mm Tip; 4 in. Length; .35mm Tip Width
Catalog No. NC9954786 Supplier Roboz Surgical Supplier No. RS5068
McPherson Forceps; Straight; Smooth Tying Platform; 0.3 X 5mm Tip; 4 in. Length; .35mm Tip Width