Learn More
Roboz Surgical MCPHERSON-VANNAS 6X.3MM STR
SDP
Catalog No. NC9686610
Encompass
McPherson-Vannas Micro Dissecting Spring Scissors; Straight; 6mm Cutting Edge; 0.15mm Tip Width; 4 in. Overall Length
Catalog No. NC9686610 Supplier Roboz Surgical Supplier No. RS5630
McPherson-Vannas Micro Dissecting Spring Scissors; Straight; 6mm Cutting Edge; 0.15mm Tip Width; 4 in. Overall Length