Learn More
Roboz Surgical Roboz Surgical McPherson Vannas Spring Scissors, Titanium, Straight, 1 ea
SDP
Catalog No. 5000574644
Encompass
These McPherson-Vannas spring scissors are precision-engineered for microsurgical applications, featuring straight blades with a 6 mm cutting edge and 0.15 mm tips. Constructed from titanium, these 4-inch instruments provide an exceptional strength-to-weight ratio to minimize hand fatigue during intricate, repetitive procedures.
  • Non-magnetic composition for safe use around strong magnetic fields and retraction systems
  • High corrosion resistance allows for use in challenging environments like saline solutions
  • Lightweight design enhances tactile feedback and surgical accuracy
  • Special blue finish reduces microscope glare and allows for quick identification
Catalog No. 50-005-74644 Supplier Roboz Surgical Supplier No. RS5630T
These McPherson-Vannas spring scissors are precision-engineered for microsurgical applications, featuring straight blades with a 6 mm cutting edge and 0.15 mm tips. Constructed from titanium, these 4-inch instruments provide an exceptional strength-to-weight ratio to minimize hand fatigue during intricate, repetitive procedures.
  • Non-magnetic composition for safe use around strong magnetic fields and retraction systems
  • High corrosion resistance allows for use in challenging environments like saline solutions
  • Lightweight design enhances tactile feedback and surgical accuracy
  • Special blue finish reduces microscope glare and allows for quick identification