Learn More
Roboz Surgical MCPHERSON-VANNAS CURVED SHAR
SDP
Supplier:  Roboz Surgical RS5605
Encompass
McPherson-Vannas Micro Dissecting Spring Scissors; Straight, Sharp Needle Points; 3mm Cutting Edge; 0.15mm Tip Width; 3 in. Overall Length These fine micro dissecting scissors have very thin blades with long taper and sharp needle points. The th
Catalog No. NC9012086