Learn More
Roboz Surgical McPherson-Vannas Micro Dissecting Spring Scissors
SDP
Catalog No. NC1438699
Encompass
McPherson-Vannas Micro Dissecting Spring Scissors; Angled, Sharp Points; 5mm Cutting Edge; 0.1mm Tip Width; 3" Overall Length.
This item is not returnable. View return policy
Catalog No. NC1438699 Supplier Roboz Surgical Supplier No. RS5606
McPherson-Vannas Micro Dissecting Spring Scissors; Angled, Sharp Points; 5mm Cutting Edge; 0.1mm Tip Width; 3" Overall Length.