Learn More
Roboz Surgical MCPHERSON-VANNAS MICRODISSECTI
SDP
Catalog No. NC9212447
Encompass
McPherson-Vannas Micro Dissecting Spring Scissors; Curved, Sharp Points; 5mm Cutting Edge; 0.1mm Tip Width; 3 in. Overall Length
Catalog No. NC9212447 Supplier Roboz Surgical Supplier No. RS5603
McPherson-Vannas Micro Dissecting Spring Scissors; Curved, Sharp Points; 5mm Cutting Edge; 0.1mm Tip Width; 3 in. Overall Length