Learn More
Roboz Surgical MCPHERSON VANNAS SCISSORS
SDP
Catalog No. NC9448531
Encompass
McPherson-Vannas Micro Dissecting Spring Scissors; Curved; 6mm Cutting Edge; 0.15mm Tip Width; 4 in. Overall Length
Catalog No. NC9448531 Supplier Roboz Surgical Supplier No. RS5631
McPherson-Vannas Micro Dissecting Spring Scissors; Curved; 6mm Cutting Edge; 0.15mm Tip Width; 4 in. Overall Length