Learn More
World Precision Instrument McPherson-Vannas Spring Scissors, 8cm, Angled, 5mm Blades
SDP
Catalog No. 502521294
Encompass
The McPherson-Vannas spring scissors were originally designed by Dr. David McPherson and Dr. Erkki Vannas in the 1970s specifically for microsurgery, which requires precision in the dissection of delicate tissues. These surgical scissors are 8cm long. The fine (0.3mm), rounded tips of these Vannas spring scissors reduce the risk of accidental tissue damage because they won't puncture. They also offer greater visibility of the surgical site in confined spaces. For even greater visibility at the tip, select the curved blades. They are available with blades that are straight, curved, angled at 45 degrees, or angled on the flat face. These surgical scissors have a smooth cutting action for a clean cut with minimal damage to surrounding tissues. The lightweight and ergonomic design of the McPherson-Vannas spring scissors reduces hand fatigue during use.
Catalog No. 50-252-1294 Supplier World Precision Instrument Supplier No. 503216
The McPherson-Vannas spring scissors were originally designed by Dr. David McPherson and Dr. Erkki Vannas in the 1970s specifically for microsurgery, which requires precision in the dissection of delicate tissues. These surgical scissors are 8cm long. The fine (0.3mm), rounded tips of these Vannas spring scissors reduce the risk of accidental tissue damage because they won't puncture. They also offer greater visibility of the surgical site in confined spaces. For even greater visibility at the tip, select the curved blades. They are available with blades that are straight, curved, angled at 45 degrees, or angled on the flat face. These surgical scissors have a smooth cutting action for a clean cut with minimal damage to surrounding tissues. The lightweight and ergonomic design of the McPherson-Vannas spring scissors reduces hand fatigue during use.