Learn More
GENEPATH DIAGNOSTICS INC MCR-1 quantitative standards for assay calibration and validation. Provided at high (10,000 copies/µL), medium (1,000 copies/µL), and low (100 copies/µL) concentrations (RUO)
Catalog No. NC3949390
Encompass_Preferred
Synthetic, sequence-verified quantitative standards targeting the mcr-1 gene associated with plasmid-mediated colistin resistance. Supplied at defined concentrations of High – 10,000 copies/µL, Medium – 1,000 copies/µL, and Low – 100 copies/µL, these standards are designed for accurate assay calibration and quantitative performance assessment across a broad dynamic range. They enable robust assay validation, sensitivity and limit-of-detection (LoD) determination, and evaluation of assay linearity in molecular assays. Compatible with real-time PCR and other nucleic acid amplification platforms, the standards ensure consistent inter-run comparability, reliable quantification, and ongoing performance monitoring. Suitable for research, assay development, and routine quality control applications, they support reliable detection and surveillance of clinically significant colistin resistance (RUO)
Catalog No. NC3949390 Supplier GENEPATH DIAGNOSTICS INC Supplier No. 10830140050
Synthetic, sequence-verified quantitative standards targeting the mcr-1 gene associated with plasmid-mediated colistin resistance. Supplied at defined concentrations of High – 10,000 copies/µL, Medium – 1,000 copies/µL, and Low – 100 copies/µL, these standards are designed for accurate assay calibration and quantitative performance assessment across a broad dynamic range. They enable robust assay validation, sensitivity and limit-of-detection (LoD) determination, and evaluation of assay linearity in molecular assays. Compatible with real-time PCR and other nucleic acid amplification platforms, the standards ensure consistent inter-run comparability, reliable quantification, and ongoing performance monitoring. Suitable for research, assay development, and routine quality control applications, they support reliable detection and surveillance of clinically significant colistin resistance (RUO)