Learn More
MCR Safety Anti-fog Perforated Goggles
Catalog No. 19808737 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Perforated; Anti-fog
Catalog No.
19-808-737 Perforated; Anti-fog
1 options
This item is not returnable. View return policy
Catalog No. 19-808-737 Supplier MCR Safety Supplier No. 2225R
This item is not returnable. View return policy
Our goggles provide a wide, unobstructed field of vision and can be worn with most half-mask respirators. The transparent protective PVC body permits a comfortable fit over prescription eyewear while protecting against impact.
  • Available in vented, perforated, and ventless styles
  • Goggle straps contain No Latex
  • Options available to protect against impact, small particles and debris
  • Rugged polycarbonate lens filters 99.9% of harmful U.V. rays
  • Exclusive Duramass™ scratch resistant coating
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1
Lens Tint Clear
Product Type Anti-fog Goggles
Ventilation Details Perforated
Material PVC