Learn More
Enterprise Technology Solutions MCR Safety Antifog Safety Glasses, UV Protection, Clear
SDP
Catalog No. 5000828205
Encompass_Preferred
Stylish Checklite Clear Antifog Safety Glasses offer exclusive Duramass scratch-resistant lens coating and flexible spatula temple design for a comfortable fit during long use. Near total seal of eye orbit assures the highest level of protection. Strong, lightweight polycarbonate lens filters out more than 99.9 percent of UV radiation. Universal nose bridge assures compliance and comfort for a variety of users. Eyewear meets ANSI Z87.1 Plus high impact.
Catalog No. 50-008-28205 Supplier Enterprise Technology Solutions Supplier No. MCSCL110AF
Stylish Checklite Clear Antifog Safety Glasses offer exclusive Duramass scratch-resistant lens coating and flexible spatula temple design for a comfortable fit during long use. Near total seal of eye orbit assures the highest level of protection. Strong, lightweight polycarbonate lens filters out more than 99.9 percent of UV radiation. Universal nose bridge assures compliance and comfort for a variety of users. Eyewear meets ANSI Z87.1 Plus high impact.