Learn More
MCR Safety BearKat™ Magnifier Safety Glasses
Catalog No. 19810335 Shop All MCR Safety Products
Change view
Click to view available options
Magnification Power:
+1.50
+2.00
+2.50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
19-810-335 +1.50
19-810-337 +2.50
19-810-336 +2.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-810-335 Supplier MCR Safety Supplier No. BKH15
Add to Cart
Add to Cart
This item is not returnable. View return policy
BearKat™ Magnifier's well thought-out construction combines the best in safety, comfort and affordability. Your vision is unobstructed by the frameless one-piece wraparound lens, which by its secure fit allows you maximum safety.
Available in a variety of diopter strengths so there is an option that is just right for you.
  • Comfort is enhanced by the flexible temples and the non-slip temple sleeves are notched on the end to accept a clip-on breakaway cord
  • 1.0, 1.5, 2.0, 2.5 Diopter available
  • Single lens design with integral side shields for unobstructed view
  • Non-Slip soft temple material
  • Exclusive Duramass scratch resistant coating
  • Rugged polycarbonate lens filters 99.9% of harmful U.V. rays
  • ANSI Z87+ and CSA Z94.3
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type Magnifier Safety Glasses
Magnification Power +1.50
Lens Material Polycarbonate
UV Protection UV400