Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety 5435 Flock-Lined Neoprene Work Gloves - 12 in.

Catalog No. 19097480D Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-097-480D X-Large
19-82-3868 Medium
19-823-867 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-097-480D Supplier MCR Safety Supplier No. 5435XL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thick, waterproof neoprene gloves are an effective replacement for similar latex or vinyl products. A textured palm enhances grip capabilities, while the extended cuff provides additional forearm protection.

  • Neoprene coating provides protection from a wide range of chemicals
  • An effective replacement for latex or vinyl gloves
  • Textured palm enhances grip capabilities
  • Maintains flexibility over a wide range of temperatures
  • Long, straight cuff for additional protection
  • Component materials comply with all FDA regulations for food contact

Specifications

Certifications/Compliance 21 CFR, 170-199
Coated Area Full Glove
Coating Material Neoprene
Color Black
Exterior Finish Partially Textured
Grade Industrial
Hand Ambidextrous
Latex Free Yes
Material Neoprene
Product Type Dip-coated Gloves
Size X-Large
Thickness (English) 30 mil
For Use With (Application) Agriculture, Construction, Oil/Gas, Petrochemical, and Plumbing
Glove Cuff Style Straight
Grip Design Textured
Liner Material Flocked Cotton
Resistant To Chemicals, Water
Series 5435
Thumb Style Straight
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.