Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Blue Beast™ Jersey-Lined Select Side-Split Leather Welding Gloves

Catalog No. 19042132 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Size:
X-Large
2X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-042132 X-Large
19-813-586 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-042132 Supplier MCR Safety Supplier No. 4600
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reinforced palm and thumb areas provide durability, with fully welted Aramid Fiber seams for increased strength and heat protection.

  • One piece, select side split cow leather back
  • Sewn with Aramid Fiber for durability
  • Soft Jersey Lining increases comfort and heat protection
  • Reinforced palm and thumb strap

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Uncoated
Color Blue
Material Cowhide Leather
Size X-Large
Style Gunn
For Use With (Application) Heliarc, Construction, and Welding
Glove Cuff Style Gauntlet
Liner Material Cotton Jersey
Series 4600
Length (English) 13.6 in.
Length (English) Cuff 5.3 in.
Length (Metric) 34.6 cm
Length (Metric) Cuff 13 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.