Learn More
MSC MCR Safety Butyl Gloves, 14 mil, Smooth, 1 Pair
Catalog No. 5000802540
Encompass
The CP14 is a 14-inch butyl rubber glove designed to provide excellent protection against harsh chemicals, including Skydrol, for aviation maintenance, mechanics, and industrial applications. Its unique solution-dipped process ensures a tight molecular structure for superior chemical impermeability.
  • Features 14 mil thickness with smooth finish
  • Gauntlet rolled cuff for extended protection
  • Excellent resistance to most acids, aldehydes, alcohols, amides, bases, esters, and ketones
  • Latex-free and anti-static properties
  • ESD-safe and TAA compliant
  • FDA accepted materials, suitable for food contact
Catalog No. 50-008-02540 Supplier MSC Supplier No. 70939368
The CP14 is a 14-inch butyl rubber glove designed to provide excellent protection against harsh chemicals, including Skydrol, for aviation maintenance, mechanics, and industrial applications. Its unique solution-dipped process ensures a tight molecular structure for superior chemical impermeability.
  • Features 14 mil thickness with smooth finish
  • Gauntlet rolled cuff for extended protection
  • Excellent resistance to most acids, aldehydes, alcohols, amides, bases, esters, and ketones
  • Latex-free and anti-static properties
  • ESD-safe and TAA compliant
  • FDA accepted materials, suitable for food contact