Learn More
MSC MCR Safety Checklite, Blue Mirror, Duramass, Small, 1 pr
Catalog No. 5000787835
Encompass
The Checklite CL1 Safety Glasses are designed for comfort, durability, and everyday protection. They feature a lightweight design and rugged polycarbonate lenses with an exclusive Duramass scratch-resistant coating, filtering 99.9% of harmful UV rays. A smaller frame ensures a secure fit for smaller face sizes.
  • Lightweight (22.9 grams) for long term wear
  • Smaller frame sized to fit smaller faces
  • Excellent orbital seal and fit
  • Provides lightweight, easy-to-wear protection
  • Rugged polycarbonate lens filters 99.9% of harmful UV rays
  • Meets ANSI/ISEA Z87.1-2025 standard
  • Exclusive Duramass scratch resistant coating
Catalog No. 50-007-87835 Supplier MSC Supplier No. 41057498
The Checklite CL1 Safety Glasses are designed for comfort, durability, and everyday protection. They feature a lightweight design and rugged polycarbonate lenses with an exclusive Duramass scratch-resistant coating, filtering 99.9% of harmful UV rays. A smaller frame ensures a secure fit for smaller face sizes.
  • Lightweight (22.9 grams) for long term wear
  • Smaller frame sized to fit smaller faces
  • Excellent orbital seal and fit
  • Provides lightweight, easy-to-wear protection
  • Rugged polycarbonate lens filters 99.9% of harmful UV rays
  • Meets ANSI/ISEA Z87.1-2025 standard
  • Exclusive Duramass scratch resistant coating