Learn More
MSC MCR Safety Checklite CL1 Glasses, Blue Mirror Lens, 1 ea
Catalog No. 5000792219
Encompass
MCR Safety Checklite CL1 Series Safety Glasses feature a lightweight design with a blue mirror polycarbonate lens, providing excellent UV protection and scratch resistance. These glasses are engineered for a secure fit and comfort during extended wear.
  • Rugged polycarbonate lens material
  • Duramass® scratch-resistant coating
  • Filters 99.9% of harmful UV rays
  • Lightweight design, less than 23 grams
  • Excellent orbital seal, sized for smaller faces
  • ANSI/ISEA Z87.1-2025 certified
  • Spatula temple type
  • CAN/CSA Z94.3, MIL-PRF-32423, TAA Compliant, Eco-Friendly MFG, AS/NZS 1337.1, CE EN166 standards met
Catalog No. 50-007-92219 Supplier MSC Supplier No. 94632601
MCR Safety Checklite CL1 Series Safety Glasses feature a lightweight design with a blue mirror polycarbonate lens, providing excellent UV protection and scratch resistance. These glasses are engineered for a secure fit and comfort during extended wear.
  • Rugged polycarbonate lens material
  • Duramass® scratch-resistant coating
  • Filters 99.9% of harmful UV rays
  • Lightweight design, less than 23 grams
  • Excellent orbital seal, sized for smaller faces
  • ANSI/ISEA Z87.1-2025 certified
  • Spatula temple type
  • CAN/CSA Z94.3, MIL-PRF-32423, TAA Compliant, Eco-Friendly MFG, AS/NZS 1337.1, CE EN166 standards met