Learn More
MSC MCR Safety Chemical Resistant Gloves, 3X-Large, 28 mil, 1 Pair
Catalog No. 5000685854
Encompass
These chemical resistant gloves are constructed with a neoprene blend over natural latex, offering robust protection for various industrial applications. With a 28 mil thickness and flock lining, they provide comfort and durability, along with excellent grip for handling diverse materials.
  • Flock lining enhances user comfort
  • Recessed diamond embossed grip improves dexterity and handling
  • Gauntlet cuff provides extended wrist and forearm protection
  • Component materials comply with FDA regulations for food contact
  • Ideal for use in food service, automotive, and general industrial settings
Catalog No. 50-006-85854 Supplier MSC Supplier No. 80111818
These chemical resistant gloves are constructed with a neoprene blend over natural latex, offering robust protection for various industrial applications. With a 28 mil thickness and flock lining, they provide comfort and durability, along with excellent grip for handling diverse materials.
  • Flock lining enhances user comfort
  • Recessed diamond embossed grip improves dexterity and handling
  • Gauntlet cuff provides extended wrist and forearm protection
  • Component materials comply with FDA regulations for food contact
  • Ideal for use in food service, automotive, and general industrial settings