Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Select Grade Unlined Grain Pigskin Leather Drivers Gloves

Catalog No. 19086296 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Series:
3400
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Series
19-086296 3400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-086296 Supplier MCR Safety Supplier No. 3400L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pigskin gloves provide abrasion resistance and breathability. Gloves become softer with use, and inherently retain their natural softness after exposure to water.

  • Select Grade Grain Pigskin
  • Straight Thumb
  • Shirred Elastic Back
  • Color-coded hem to identify size

Specifications

ANSI Abrasion Rating ANSI 3
Color Beige
Grade Select
Hand Specific
Material Pigskin Leather
Product Type Driver Gloves
Size Large
Style Gunn
For Use With (Application) Carpentry, Farming, Landscaping, Transportation, and Heavy Equipment Operation
Glove Cuff Style Open
Hem Color Brown
Lining Unlined
Packaging Type Polybag
Series 3400
Thumb Style Straight
Length (English) 9.8 in.
Length (Metric) 24.8 cm
Show More Show Less

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.