Learn More
MCR Safety Crews™ Storm™ Safety Glasses
Catalog No. 19003047 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Frame Color:
Black
Navy
Lens Coating:
Scratch-resistant
Lens Tint:
Clear
Gray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
19-003-047 Navy Scratch-resistant Clear
19-003-044 Black Scratch-resistant Clear
19-003-045 Black - Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-003-047 Supplier MCR Safety Supplier No. ST120
This item is not returnable. View return policy
Workers in the Storm™ can expect extreme comfort with this face-flattering protective eyewear from Crews™. Ratchet action temples combined with non-slip rubber head grips create a customized fit for virtually any head size.
  • Available in a variety of frame colors and lens combinations
  • Wraparound lens constructed of lightweight, scratch resistant polycarbonate, that filters 99.9% of U.V. rays, delivers advanced visual acuity and protection
  • Single wraparound dual focal point lens with integral side shields
  • 5 position ratchet action bayonet temples
  • Exclusive Duramass™ scratch resistant coating
  • Non-slip, soft temple material
Compliance
Spectacles meet ANSI Z87.1-1989 standards for optical clarity and impact resistance.
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance Spectacles meet ANSI Z87.1-1989 standards for optical clarity and impact resistance.
Lens Tint Clear
Frame Color Navy
Lens Material Polycarbonate