Learn More
MSC MCR Safety Cut-Resistant Gloves, Ninja Max, Dyneema Bi-Polymer, 1 Pair
Catalog No. 5000675616
Encompass
These gloves feature a 10-gauge Dyneema® Diamond Technology and synthetic shell with a bi-polymer coating on the palm and fingertips. They offer cut and abrasion resistance, providing a thinner and lighter design with exceptional protection and durability.
  • Features a 10-gauge Dyneema®/synthetic shell
  • Bi-polymer coating on palm and fingertips
  • Cut-resistant and abrasion-resistant
  • Point of purchase hang tag
  • Treated with Actifresh®
  • Touchscreen compatible
  • Eco-friendly packaging
Catalog No. 50-006-75616 Supplier MSC Supplier No. 91609446
These gloves feature a 10-gauge Dyneema® Diamond Technology and synthetic shell with a bi-polymer coating on the palm and fingertips. They offer cut and abrasion resistance, providing a thinner and lighter design with exceptional protection and durability.
  • Features a 10-gauge Dyneema®/synthetic shell
  • Bi-polymer coating on palm and fingertips
  • Cut-resistant and abrasion-resistant
  • Point of purchase hang tag
  • Treated with Actifresh®
  • Touchscreen compatible
  • Eco-friendly packaging