Learn More
MSC MCR Safety Cut-Resistant Work Gloves, 10-Gauge Dyneema Diamond, 1 Pair
Catalog No. 5000686730
Encompass
The N9676G features a 10-gauge Dyneema Diamond Technology and synthetic shell with a bi-polymer coating on the palm and fingertips. This bi-polymer coated glove offers cut and abrasion resistance and provides great gripping power. Dyneema Diamond Technology fiber is 40% lighter (less dense) than aramid fiber, resulting in gloves that are thinner and lighter while offering exceptional cut protection. It provides cool-touch comfort and maximum durability.
  • Shell made of 10-gauge Dyneema Diamond Technology and synthetic material
  • Bi-polymer coating on palm and fingertips
  • Cut-resistant and abrasion-resistant gloves
  • Point of purchase hang tag
  • Treated with Actifresh
  • Cool-touch comfort
  • Touchscreen compatible
  • Knit wrist cuff style
Catalog No. 50-006-86730 Supplier MSC Supplier No. 12872099
The N9676G features a 10-gauge Dyneema Diamond Technology and synthetic shell with a bi-polymer coating on the palm and fingertips. This bi-polymer coated glove offers cut and abrasion resistance and provides great gripping power. Dyneema Diamond Technology fiber is 40% lighter (less dense) than aramid fiber, resulting in gloves that are thinner and lighter while offering exceptional cut protection. It provides cool-touch comfort and maximum durability.
  • Shell made of 10-gauge Dyneema Diamond Technology and synthetic material
  • Bi-polymer coating on palm and fingertips
  • Cut-resistant and abrasion-resistant gloves
  • Point of purchase hang tag
  • Treated with Actifresh
  • Cool-touch comfort
  • Touchscreen compatible
  • Knit wrist cuff style