Learn More
MSC MCR Safety DBL Matrix Face Shield Window, Polycarbonate, 1 ea
Catalog No. 5000686234
Encompass
This replacement face shield window is constructed from durable polycarbonate, designed to provide clear vision and reliable protection in a variety of industrial and laboratory settings. Its double matrix design ensures optical clarity and robust performance.
  • Made of polycarbonate
  • Features a double matrix design
  • Measures 7 x 16.75 x 0.040 in
  • Provides protection for face shields
Catalog No. 50-006-86234 Supplier MSC Supplier No. 64104714
This replacement face shield window is constructed from durable polycarbonate, designed to provide clear vision and reliable protection in a variety of industrial and laboratory settings. Its double matrix design ensures optical clarity and robust performance.
  • Made of polycarbonate
  • Features a double matrix design
  • Measures 7 x 16.75 x 0.040 in
  • Provides protection for face shields