Learn More
MSC MCR Safety Disposable Gloves, Latex, Powder-Free, 5 mil, Large, 100/Pk.
Catalog No. 5000803459
Encompass
These SensaTouch™ gloves are crafted from natural rubber latex, offering a 5 mil thickness and a textured finish. Designed for industrial and food service applications, they are powder-free and biodegradable, with materials that comply with federal regulations for food contact.
  • Thickness of 5 mil (4.9 g)
  • Powder-free and disposable
  • Industrial food service grade
  • Textured finish for enhanced grip
  • Overall length of 9.5 in.
  • Natural color
  • Ambidextrous design
  • Rolled cuff style
  • Inherently antimicrobial and antibacterial
  • FDA accepted material
Catalog No. 50-008-03459 Supplier MSC Supplier No. 71299622
These SensaTouch™ gloves are crafted from natural rubber latex, offering a 5 mil thickness and a textured finish. Designed for industrial and food service applications, they are powder-free and biodegradable, with materials that comply with federal regulations for food contact.
  • Thickness of 5 mil (4.9 g)
  • Powder-free and disposable
  • Industrial food service grade
  • Textured finish for enhanced grip
  • Overall length of 9.5 in.
  • Natural color
  • Ambidextrous design
  • Rolled cuff style
  • Inherently antimicrobial and antibacterial
  • FDA accepted material