Learn More
MSC MCR Safety Disposable Nitrile Gloves, 6 mil, Powder Free, 100/dispenser
Catalog No. 5000684076
Encompass
These gloves feature a patented suction grip design that works effectively with most wet and greasy surfaces. An embossed internal grip provides additional traction, preventing the gloves from slipping against the skin. They are suitable for various industrial uses, including automotive, assembly, paint spraying, electro-plating, cleaning, asbestos removal, and janitorial work.
  • Nitrile, 6 mil (6.2 g)
  • Powder free disposable nitrile
  • Industrial food service grade
  • Textured grip
  • Overall length of 9.5 in.
  • Orange color
  • Passes ASTM D6978 for fentanyl handling
  • Suitable for gastric acid fluid contact
  • Fish-scale pattern texture on both sides and inner thumb
  • Touchscreen compatible
  • FDA accepted material
  • Ambidextrous hand orientation
  • Rolled cuff style
  • Wing thumb style
Catalog No. 50-006-84076 Supplier MSC Supplier No. 37904497
These gloves feature a patented suction grip design that works effectively with most wet and greasy surfaces. An embossed internal grip provides additional traction, preventing the gloves from slipping against the skin. They are suitable for various industrial uses, including automotive, assembly, paint spraying, electro-plating, cleaning, asbestos removal, and janitorial work.
  • Nitrile, 6 mil (6.2 g)
  • Powder free disposable nitrile
  • Industrial food service grade
  • Textured grip
  • Overall length of 9.5 in.
  • Orange color
  • Passes ASTM D6978 for fentanyl handling
  • Suitable for gastric acid fluid contact
  • Fish-scale pattern texture on both sides and inner thumb
  • Touchscreen compatible
  • FDA accepted material
  • Ambidextrous hand orientation
  • Rolled cuff style
  • Wing thumb style