Learn More
Enterprise Technology Solutions MCR Safety Economy Goggle, Wide, Perforated Frame, PVC Body, CL
SDP
Catalog No. 5000828232
Encompass_Preferred
Economy goggles protect eyes from flying particles and can be worn with most half-mask respirators. The transparent, protective PVC body permits a comfortable fit over prescription eyewear while protecting against impact. Design features hooded indirect ventilation for perforations along the top and sides, flexible frame, and large, polycarbonate lens for an unobstructed view. Goggles meet ANSI Z87.1-2003 High-impact standards.
Catalog No. 50-008-28232 Supplier Enterprise Technology Solutions Supplier No. MCS2220
Economy goggles protect eyes from flying particles and can be worn with most half-mask respirators. The transparent, protective PVC body permits a comfortable fit over prescription eyewear while protecting against impact. Design features hooded indirect ventilation for perforations along the top and sides, flexible frame, and large, polycarbonate lens for an unobstructed view. Goggles meet ANSI Z87.1-2003 High-impact standards.