Learn More
Enterprise Technology Solutions MCR Safety Eyewear,Adjustable Temples,11 Degree Base,Matte Black Frame
SDP
Catalog No. 5000828202
Encompass_Preferred
Safety glasses feature a matte black frame finish, adjustable temples, 11-degree base curve lens and proprietary gel nosepiece. Scratch-resistant lens is made of strong, lightweight polycarbonate. Frame and temple are made of nylon. Nosepiece is made of gel for extra comfort. Glasses filter 99.9 percent of UV radiation and meet or exceed ANSI Z87.1 safety standards.
Catalog No. 50-008-28202 Supplier Enterprise Technology Solutions Supplier No. MCSKD110
Safety glasses feature a matte black frame finish, adjustable temples, 11-degree base curve lens and proprietary gel nosepiece. Scratch-resistant lens is made of strong, lightweight polycarbonate. Frame and temple are made of nylon. Nosepiece is made of gel for extra comfort. Glasses filter 99.9 percent of UV radiation and meet or exceed ANSI Z87.1 safety standards.