Description
- Engineered to provide the highest level of innovation for consumers that demand the very best in safety
- Meets military MIL-PRF-31013 clause 3.5.1.1 for high velocity impact protection and exceed ANSI Z87+
- Patented multi-injection molding of the lens and frame
- Allows the frame to flex and fit faces from large to small
- Exclusive Duramass™ scratch resistant coating
- Rugged polycarbonate lens filters 99.9% of harmful U.V. rays
Specifications
Specifications
| Lens Tint | Clear |
| Frame Color | Translucent Brown |
| Frame Material | Flexible Thermo Plastic Urethane |