Learn More
MSC MCR Safety General Purpose Gloves, Cowhide Leather, X-Large, 1 Pair
Catalog No. 5000788490
Encompass
These general purpose gloves are constructed from select grade grain cowhide leather, featuring a double palm and wing thumb design for enhanced durability and comfort. They are ideal for a wide range of general applications requiring robust abrasion protection.
  • Cowhide leather lining and back material
  • Slip-on cuff style for easy wear
  • X-Large size with numeric size 10
  • Brown and beige color scheme
  • ANSI/ISEA abrasion resistance level 4
  • Full fingered design with wing thumb
  • Double palm for enhanced durability
  • Color-coded hem for size identification
Catalog No. 50-007-88490 Supplier MSC Supplier No. 40744252
These general purpose gloves are constructed from select grade grain cowhide leather, featuring a double palm and wing thumb design for enhanced durability and comfort. They are ideal for a wide range of general applications requiring robust abrasion protection.
  • Cowhide leather lining and back material
  • Slip-on cuff style for easy wear
  • X-Large size with numeric size 10
  • Brown and beige color scheme
  • ANSI/ISEA abrasion resistance level 4
  • Full fingered design with wing thumb
  • Double palm for enhanced durability
  • Color-coded hem for size identification