Learn More
MSC MCR Safety General Purpose Gloves, Fiber-Lined, FDA Approved, 1 Pair
Catalog No. 5000688738
Encompass
MCR Safety General Purpose Gloves are fiber-lined gloves designed for general purpose applications. These white, 8 in. long gloves are FDA approved and suitable for various tasks requiring hand protection.
  • Component materials comply with all federal regulations for food contact
  • Fast evaporation rate of moisture
  • Greater thermal insulation
  • One size
  • Hollow-core hydrophobic polyester fiber
  • Knit wrist cuff style
  • Full fingered coverage
  • Unisex gender
Catalog No. 50-006-88738 Supplier MSC Supplier No. 93468759
MCR Safety General Purpose Gloves are fiber-lined gloves designed for general purpose applications. These white, 8 in. long gloves are FDA approved and suitable for various tasks requiring hand protection.
  • Component materials comply with all federal regulations for food contact
  • Fast evaporation rate of moisture
  • Greater thermal insulation
  • One size
  • Hollow-core hydrophobic polyester fiber
  • Knit wrist cuff style
  • Full fingered coverage
  • Unisex gender