Learn More
MSC MCR Safety General Purpose Gloves, Goatskin Leather, Large, 1 Pair
Catalog No. 5000796123
Encompass
These MCR Safety general purpose gloves are constructed from goatskin leather, designed for various applications requiring hand protection. They feature a padded palm grip for enhanced comfort and dexterity during use.
  • Level 1 impact rating
  • Shirred elastic back for a secure fit
  • Supple grain goatskin is soft and comfortable
  • TPR back for added impact protection
  • Keystone thumb type
  • Slip-on cuff style
  • Provides protection against impact, abrasion, heat, and puncture
Catalog No. 50-007-96123 Supplier MSC Supplier No. 19940543
These MCR Safety general purpose gloves are constructed from goatskin leather, designed for various applications requiring hand protection. They feature a padded palm grip for enhanced comfort and dexterity during use.
  • Level 1 impact rating
  • Shirred elastic back for a secure fit
  • Supple grain goatskin is soft and comfortable
  • TPR back for added impact protection
  • Keystone thumb type
  • Slip-on cuff style
  • Provides protection against impact, abrasion, heat, and puncture