Learn More
MSC MCR Safety General Purpose Gloves, Large, Cowhide Leather, 1 Pair
Catalog No. 5000768165
Encompass
These general purpose gloves are designed for various applications, offering a smooth grip and a large size. They are constructed with a cowhide leather lining and a fabric back, featuring a rubberized gauntlet cuff for enhanced safety and comfort. These full-fingered gloves provide protection against abrasion, heat, cut, and puncture.
  • Cowhide leather lining and palm
  • Fabric back
  • Rubberized gauntlet cuff, 4.5 in safety cuff
  • Smooth grip surface
  • Large size (numeric size 9)
  • Wing thumb type
  • Overall length: 13-3/16 in
  • ANSI/ISEA abrasion resistance level 4
  • Provides protection against abrasion, heat, cut, and puncture
  • Maximum operating temperature: 392 degrees Fahrenheit
Catalog No. 50-007-68165 Supplier MSC Supplier No. 40744187
These general purpose gloves are designed for various applications, offering a smooth grip and a large size. They are constructed with a cowhide leather lining and a fabric back, featuring a rubberized gauntlet cuff for enhanced safety and comfort. These full-fingered gloves provide protection against abrasion, heat, cut, and puncture.
  • Cowhide leather lining and palm
  • Fabric back
  • Rubberized gauntlet cuff, 4.5 in safety cuff
  • Smooth grip surface
  • Large size (numeric size 9)
  • Wing thumb type
  • Overall length: 13-3/16 in
  • ANSI/ISEA abrasion resistance level 4
  • Provides protection against abrasion, heat, cut, and puncture
  • Maximum operating temperature: 392 degrees Fahrenheit