Learn More
MSC MCR Safety Glasses, Polycarbonate, Clear Mirror, ANSI Z87.1, 1 ea
Catalog No. 5000788635
Encompass
These safety glasses feature scratch-resistant, polycarbonate, clear mirror lenses and a half-framed design. They have a black frame with adjustable temples and meet ANSI Z87.1+ standards, making them suitable for indoor/outdoor applications.
  • Scratch-resistant polycarbonate lenses
  • Clear mirror lens color
  • Half-framed, wrap-around design
  • Black frame with adjustable bayonet temples
  • Universal size
  • UV protection
  • Meets ANSI Z87.1+ standards
Catalog No. 50-007-88635 Supplier MSC Supplier No. 01892975
These safety glasses feature scratch-resistant, polycarbonate, clear mirror lenses and a half-framed design. They have a black frame with adjustable temples and meet ANSI Z87.1+ standards, making them suitable for indoor/outdoor applications.
  • Scratch-resistant polycarbonate lenses
  • Clear mirror lens color
  • Half-framed, wrap-around design
  • Black frame with adjustable bayonet temples
  • Universal size
  • UV protection
  • Meets ANSI Z87.1+ standards