Learn More
MSC MCR Safety Glasses, Scratch Resistant, Blue Mirror, Full-Framed, 1 ea
Catalog No. 5000803296
Encompass
These safety glasses are designed for protection during everyday operations. They feature a full-framed design with scratch-resistant polycarbonate blue mirror lenses and meet ANSI Z87.1+ standards.
  • Scratch resistant coatings prevent damage from minor debris
  • Full-framed design with black frame
  • Adjustable temples
  • Polycarbonate blue mirror lenses
  • UV protection
  • Adjustable nose pads and arms
  • Meets ANSI Z87.1+ standards
Catalog No. 50-008-03296 Supplier MSC Supplier No. 33881673
These safety glasses are designed for protection during everyday operations. They feature a full-framed design with scratch-resistant polycarbonate blue mirror lenses and meet ANSI Z87.1+ standards.
  • Scratch resistant coatings prevent damage from minor debris
  • Full-framed design with black frame
  • Adjustable temples
  • Polycarbonate blue mirror lenses
  • UV protection
  • Adjustable nose pads and arms
  • Meets ANSI Z87.1+ standards