Learn More
MSC MCR Safety Glasses, Scratch-Resistant Polycarbonate Lens, 2.00 Diopter, 1 ea
Catalog No. 5000796523
Encompass
These safety glasses feature scratch-resistant, polycarbonate, clear lenses with a black frame and non-adjustable temples. They meet ANSI Z87.1+ standards, offer UV protection, and include a 2.00 diopter lens.
  • Scratch-resistant lens coating
  • Polycarbonate lens and frame material
  • Clear lens color
  • Black frame color
  • Non-adjustable temples
  • UV protection
  • ANSI Z87.1+ compliant
  • Bayonet temple type
  • Includes cord
Catalog No. 50-007-96523 Supplier MSC Supplier No. 14220560
These safety glasses feature scratch-resistant, polycarbonate, clear lenses with a black frame and non-adjustable temples. They meet ANSI Z87.1+ standards, offer UV protection, and include a 2.00 diopter lens.
  • Scratch-resistant lens coating
  • Polycarbonate lens and frame material
  • Clear lens color
  • Black frame color
  • Non-adjustable temples
  • UV protection
  • ANSI Z87.1+ compliant
  • Bayonet temple type
  • Includes cord