Learn More
MSC MCR Safety Gloves, 7 Gauge Dotted Cotton/Poly, Small, 12 Pairs
Catalog No. 5000674534
Encompass
The 9660SM Dotted Cotton String Knit Work Gloves offer general-purpose hand protection, providing grip and light abrasion resistance. Constructed from a comfortable 7 gauge cotton/polyester blend, these gloves are breathable and help prevent minor cuts and scrapes.
  • Pvc dots on both sides enhance grip and abrasion resistance, extending glove lifespan.
  • Seven gauge regular weight string knit construction ensures excellent breathability for extended wear.
  • Reversible design with pvc dots on both sides doubles usable life for increased efficiency.
  • Color-coded hem assists in size identification.
  • Comfortable, soft, and breathable for all-day use.
Catalog No. 50-006-74534 Supplier MSC Supplier No. 52343886
The 9660SM Dotted Cotton String Knit Work Gloves offer general-purpose hand protection, providing grip and light abrasion resistance. Constructed from a comfortable 7 gauge cotton/polyester blend, these gloves are breathable and help prevent minor cuts and scrapes.
  • Pvc dots on both sides enhance grip and abrasion resistance, extending glove lifespan.
  • Seven gauge regular weight string knit construction ensures excellent breathability for extended wear.
  • Reversible design with pvc dots on both sides doubles usable life for increased efficiency.
  • Color-coded hem assists in size identification.
  • Comfortable, soft, and breathable for all-day use.