Learn More
MSC MCR Safety Gloves, Cowhide, Smooth, Large, 1 ea
Catalog No. 5000686601
Encompass
MCR Safety General Purpose Gloves offer reliable hand protection for various applications. Constructed from durable cowhide leather, these gloves provide excellent strength and a smooth grip, designed to protect against abrasion, heat, and puncture.
  • Color-coded hem for size identification
  • CV grade cow grain for excellent strength
  • Keystone thumb for comfort and dexterity
  • Shirred elastic back for enhanced fit and safety
  • Slip-on cuff style
  • Smooth grip surface
  • Protection against abrasion, heat, and puncture
  • Maximum operating temperature of 284°F
Catalog No. 50-006-86601 Supplier MSC Supplier No. 52387388
MCR Safety General Purpose Gloves offer reliable hand protection for various applications. Constructed from durable cowhide leather, these gloves provide excellent strength and a smooth grip, designed to protect against abrasion, heat, and puncture.
  • Color-coded hem for size identification
  • CV grade cow grain for excellent strength
  • Keystone thumb for comfort and dexterity
  • Shirred elastic back for enhanced fit and safety
  • Slip-on cuff style
  • Smooth grip surface
  • Protection against abrasion, heat, and puncture
  • Maximum operating temperature of 284°F