Learn More
MSC MCR Safety Gloves, Cut A4, Kevlar, 1 Pair
Catalog No. 5000803729
Encompass
These gloves are constructed from durable 10-gauge Kevlar and feature a blue crinkle latex coating on the palm and fingers. They provide robust protection against cuts, punctures, and abrasion, while offering superior grip and high comfort.
  • Durable 10-gauge Kevlar construction
  • Blue crinkle latex coating on palm and fingers for enhanced grip
  • ANSI/ISEA Cut Resistance Level A4
  • ANSI/ISEA Puncture Resistance Level 5
  • ANSI/ISEA Abrasion Resistance Level 4
  • Knit style for flexibility and comfort
  • Full-fingered design with Kevlar lining
Catalog No. 50-008-03729 Supplier MSC Supplier No. 43783166
These gloves are constructed from durable 10-gauge Kevlar and feature a blue crinkle latex coating on the palm and fingers. They provide robust protection against cuts, punctures, and abrasion, while offering superior grip and high comfort.
  • Durable 10-gauge Kevlar construction
  • Blue crinkle latex coating on palm and fingers for enhanced grip
  • ANSI/ISEA Cut Resistance Level A4
  • ANSI/ISEA Puncture Resistance Level 5
  • ANSI/ISEA Abrasion Resistance Level 4
  • Knit style for flexibility and comfort
  • Full-fingered design with Kevlar lining