Learn More
MSC MCR Safety Gloves, Cut A4, Puncture 2, X-Large, 1 Pair
Catalog No. 5000803903
Encompass
These gloves provide advanced hand protection, specifically designed for environments requiring high resistance to cuts, punctures, and abrasions. Constructed with durable materials, they offer both comfort and robust defense against various workplace hazards.
  • Mechanic's style for versatile utility
  • Palm coated with silicone for enhanced grip
  • ANSI/ISEA Cut Resistance Level A4 for superior cut protection
  • ANSI/ISEA Puncture Resistance Level 2
  • ANSI/ISEA Abrasion Resistance Level 5
  • Primary material is HPPE for durability
  • HyperMax lining for additional cut resistance
  • Textured grip surface ensures secure handling
  • Thermoplastic elastomer back-of-hand protection with TPR provides impact resistance
  • Slip-on cuff style with hook and loop closure for a secure fit
  • Breathable fourchettes enhance comfort during extended wear
Catalog No. 50-008-03903 Supplier MSC Supplier No. 14220438
These gloves provide advanced hand protection, specifically designed for environments requiring high resistance to cuts, punctures, and abrasions. Constructed with durable materials, they offer both comfort and robust defense against various workplace hazards.
  • Mechanic's style for versatile utility
  • Palm coated with silicone for enhanced grip
  • ANSI/ISEA Cut Resistance Level A4 for superior cut protection
  • ANSI/ISEA Puncture Resistance Level 2
  • ANSI/ISEA Abrasion Resistance Level 5
  • Primary material is HPPE for durability
  • HyperMax lining for additional cut resistance
  • Textured grip surface ensures secure handling
  • Thermoplastic elastomer back-of-hand protection with TPR provides impact resistance
  • Slip-on cuff style with hook and loop closure for a secure fit
  • Breathable fourchettes enhance comfort during extended wear