Learn More
MSC MCR Safety Gloves, Cut A6, Puncture 4, X-Large, 1 Pair
Catalog No. 5000793223
Encompass
These gloves are designed for cut, puncture, and abrasion resistance. They are made from HPPE and feature a polyurethane coating on the palm, making them suitable for various industrial applications requiring enhanced protection.
  • Knit style
  • Polyurethane coated palm and fingertips
  • ANSI/ISEA Cut Resistance Level A6
  • HPPE primary material
  • ANSI/ISEA Abrasion Resistance Level 5
  • ANSI/ISEA Puncture Resistance Level 4
  • Seamless 13 gauge shell
  • X-Large size, 11 in overall length
  • High-visibility impact protection
  • Reinforced thumb crotch
  • Reflective cuff logo
Catalog No. 50-007-93223 Supplier MSC Supplier No. 84556026
These gloves are designed for cut, puncture, and abrasion resistance. They are made from HPPE and feature a polyurethane coating on the palm, making them suitable for various industrial applications requiring enhanced protection.
  • Knit style
  • Polyurethane coated palm and fingertips
  • ANSI/ISEA Cut Resistance Level A6
  • HPPE primary material
  • ANSI/ISEA Abrasion Resistance Level 5
  • ANSI/ISEA Puncture Resistance Level 4
  • Seamless 13 gauge shell
  • X-Large size, 11 in overall length
  • High-visibility impact protection
  • Reinforced thumb crotch
  • Reflective cuff logo